Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107534.1 | NP_001101004.1 | 6 - 7 | 574 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |