Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107321.1 | NP_001100791.1 | 2 - 3 | 715 | 59 | |
XM_008771355.1 | XP_008769577.1 | 2 - 3 | 3858 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Rattus norvegicus Probe spans exons |