Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014239.1 | NP_001014261.1 | 2 - 3 | 166 | 132 | |
GenBank mRNA | BC082088.1 | - | 2 - 3 | 166 | 7133991 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |