Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230922.2 | XP_006230984.1 | 6 - 7 | 766 | 61 | |
XM_006230923.2 | XP_006230985.1 | 6 - 7 | 1026 | 61 | ||
XM_006230924.2 | XP_006230986.1 | 6 - 7 | 823 | 61 | ||
XM_008760190.1 | XP_008758412.1 | 6 - 7 | 669 | 61 | ||
XM_574595.6 | XP_574595.3 | 6 - 7 | 1347 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |