Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012744.2 | NP_036876.2 | 11 - 12 | 1576 | 102 | |
XM_006230689.2 | XP_006230751.1 | 12 - 13 | 1807 | 102 | ||
XM_006230690.2 | XP_006230752.1 | 11 - 12 | 1922 | 102 | ||
XM_006230691.2 | XP_006230753.1 | 12 - 13 | 1629 | 102 | ||
XM_006230692.2 | XP_006230754.1 | 11 - 12 | 1599 | 102 | ||
XM_006230693.2 | XP_006230755.1 | 12 - 13 | 1731 | 102 | ||
XM_006230694.2 | XP_006230756.1 | 11 - 12 | 1598 | 102 | ||
XM_008760087.1 | XP_008758309.1 | 11 - 12 | 1704 | 102 | ||
GenBank mRNA | BC085680.1 | - | 11 - 12 | 1553 | 7109369 | 102 |
U32314.1 | - | 11 - 12 | 1544 | 102 | ||
U36585.1 | - | 11 - 12 | 1576 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |