Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022698.1 | NP_073189.1 | 1 - 2 | 118 | 112 | |
XM_006230896.2 | XP_006230958.1 | 1 - 2 | 823 | 112 | ||
GenBank mRNA | AF003523.1 | - | 1 - 2 | 118 | 112 | |
AF031227.1 | - | 1 - 2 | 156 | 112 | ||
AF279910.1 | - | 1 - 2 | 118 | 112 | ||
AF279911.1 | - | 1 - 2 | 118 | 112 | ||
BC061728.1 | - | 1 - 2 | 363 | 5598682 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |