Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Deaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031801.1 | NP_113989.1 | 3 - 4 | 620 | 128 | |
XM_006230582.2 | XP_006230644.1 | 3 - 4 | 1488 | 128 | ||
XM_006230583.2 | XP_006230645.1 | 3 - 4 | 1488 | 128 | ||
GenBank mRNA | AF055884.1 | - | 3 - 4 | 620 | 128 | |
U59659.1 | - | 3 - 4 | 369 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |