Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Deaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031801.1 | NP_113989.1 | 1 - 2 | 387 | 71 | |
XM_008760011.1 | XP_008758233.1 | 1 - 2 | 291 | 71 | ||
XM_008760012.1 | XP_008758234.1 | 1 - 2 | 291 | 71 | ||
XM_008760013.1 | XP_008758235.1 | 1 - 2 | 291 | 71 | ||
GenBank mRNA | AF055884.1 | - | 1 - 2 | 387 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |