Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107555.1 | NP_001101025.1 | 3 - 4 | 457 | 70 | |
XM_006230401.1 | XP_006230463.1 | 2 - 3 | 294 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |