Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efemp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005907.2 | NP_001005907.1 | 7 - 8 | 831 | 61 | |
NM_001277341.1 | NP_001264270.1 | 8 - 9 | 954 | 61 | ||
NM_001277346.1 | NP_001264275.1 | 6 - 7 | 624 | 61 | ||
XM_006230742.2 | XP_006230804.1 | 8 - 9 | 1294 | 61 | ||
XM_006230743.1 | XP_006230805.1 | 8 - 9 | 945 | 61 | ||
XM_006230744.2 | XP_006230806.1 | 8 - 9 | 951 | 61 | ||
GenBank mRNA | BC083804.1 | - | 7 - 8 | 769 | 7109182 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |