Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098241.1 | NP_001091711.1 | 4 - 5 | 623 | 106 | |
NM_001130441.1 | NP_001123913.1 | 4 - 5 | 623 | 106 | ||
XM_006230535.2 | XP_006230597.2 | 4 - 5 | 617 | 106 | ||
GenBank mRNA | BC086608.1 | - | 4 - 5 | 619 | 106 | |
BC099130.1 | - | 4 - 5 | 645 | 106 | ||
CO567790.1 | - | 4 - 5 | 623 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |