Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014167.1 | NP_001014189.1 | 3 - 4 | 581 | 72 | |
XM_006231293.1 | XP_006231355.1 | 3 - 4 | 1785 | 72 | ||
XM_006231294.1 | XP_006231356.1 | 3 - 4 | 1785 | 72 | ||
XM_008760345.1 | XP_008758567.1 | 3 - 4 | 464 | 72 | ||
GenBank mRNA | BC085833.1 | - | 3 - 4 | 581 | 7134816 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |