Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Denr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115046.2 | NP_001108518.1 | 2 - 3 | 171 | 95 | |
XM_008769260.1 | XP_008767482.1 | 2 - 3 | 222 | 95 | ||
GenBank mRNA | BC158753.1 | - | 2 - 3 | 171 | 9102903 | 95 |
FQ210696.1 | - | 2 - 3 | 193 | 95 | ||
FQ212845.1 | - | 2 - 3 | 191 | 95 | ||
FQ220357.1 | - | 2 - 3 | 228 | 95 | ||
FQ232826.1 | - | 2 - 3 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |