Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012748.1 | NP_036880.1 | 3 - 4 | 208 | 88 | |
XM_006249249.2 | XP_006249311.1 | 3 - 4 | 366 | 88 | ||
XM_006249250.2 | XP_006249312.1 | 3 - 4 | 366 | 88 | ||
XM_006249253.2 | XP_006249315.1 | 4 - 5 | 2112 | 88 | ||
XM_008769188.1 | XP_008767410.1 | 4 - 5 | 2579 | 88 | ||
GenBank mRNA | AY302700.1 | - | 3 - 4 | 208 | 88 | |
L20823.1 | - | 3 - 4 | 208 | 88 | ||
L20888.1 | - | 3 - 4 | 208 | 88 | ||
L20889.1 | - | 3 - 4 | 208 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |