Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012748.1 | NP_036880.1 | 6 - 7 | 469 | 121 | |
XM_006249249.2 | XP_006249311.1 | 6 - 7 | 627 | 121 | ||
XM_006249250.2 | XP_006249312.1 | 6 - 7 | 627 | 121 | ||
XM_006249253.2 | XP_006249315.1 | 7 - 8 | 2373 | 121 | ||
XM_008769188.1 | XP_008767410.1 | 7 - 8 | 2840 | 121 | ||
GenBank mRNA | AY302700.1 | - | 6 - 7 | 469 | 121 | |
L20823.1 | - | 6 - 7 | 469 | 121 | ||
L20888.1 | - | 6 - 7 | 469 | 121 | ||
L20889.1 | - | 6 - 7 | 469 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |