Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135840.1 | NP_001129312.1 | 6 - 7 | 883 | 67 |
Gene Symbol: LOC100912590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751210.3 | XP_003751258.2 | 963 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |