Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127570.1 | NP_001121042.1 | 3 - 4 | 511 | 78 | |
GenBank mRNA | BC161996.1 | - | 3 - 4 | 511 | 9034684 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |