Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105919.1 | NP_001099389.1 | 3 - 4 | 519 | 56 | |
XM_006249121.2 | XP_006249183.1 | 3 - 4 | 585 | 56 | ||
XM_006249122.2 | XP_006249184.1 | 3 - 4 | 539 | 56 | ||
XM_006249123.2 | XP_006249185.1 | 3 - 4 | 565 | 56 | ||
GenBank mRNA | BC167057.1 | - | 3 - 4 | 532 | 9098277 | 56 |
FQ213833.1 | - | 3 - 4 | 345 | 56 | ||
FQ222246.1 | - | 3 - 4 | 331 | 56 | ||
FQ232488.1 | - | 3 - 4 | 331 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |