Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105919.1 | NP_001099389.1 | 1 - 2 | 309 | 103 | |
XM_006249122.2 | XP_006249184.1 | 1 - 2 | 329 | 103 | ||
GenBank mRNA | BC167057.1 | - | 1 - 2 | 322 | 9098277 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |