Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053762.1 | NP_446214.1 | 2 - 3 | 462 | 54 | |
GenBank mRNA | BC127488.1 | - | 2 - 3 | 462 | 7308805 | 54 |
D78482.1 | - | 2 - 3 | 462 | 54 | ||
Y10823.1 | - | 2 - 3 | 455 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |