Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wsb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007616.1 | NP_001007617.1 | 6 - 7 | 897 | 74 | |
XM_008769220.1 | XP_008767442.1 | 5 - 6 | 834 | 74 | ||
GenBank mRNA | BC081901.1 | - | 6 - 7 | 897 | 7110808 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |