Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfip11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008291.1 | NP_001008292.1 | 3 - 4 | 482 | 80 | |
GenBank mRNA | BC085809.1 | - | 3 - 4 | 482 | 7113767 | 80 |
DQ342031.1 | - | 3 - 4 | 481 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |