Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025112.1 | NP_001020283.1 | 1 - 2 | 241 | 75 | |
GenBank mRNA | AB040671.1 | - | 1 - 2 | 241 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |