Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105929.1 | NP_001099399.1 | 11 - 12 | 1232 | 152 | |
XM_006249333.2 | XP_006249395.1 | 10 - 11 | 1206 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |