Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oasl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009681.1 | NP_001009681.1 | 3 - 4 | 659 | 88 | |
GenBank mRNA | AY227756.1 | - | 3 - 4 | 659 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |