Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC158866.1 | - | 1 - 2 | 247 | 5620924 | 61 |
FQ212189.1 | - | 1 - 2 | 260 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |