Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CB715798.1 | - | 2 - 3 | 275 | 68 | |
FQ212189.1 | - | 9 - 10 | 1350 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |