Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105933.1 | NP_001099403.1 | 5 - 6 | 481 | 72 | |
GenBank mRNA | BC166554.1 | - | 5 - 6 | 440 | 7311069 | 72 |
FQ219862.1 | - | 5 - 6 | 455 | 72 | ||
FQ221109.1 | - | 5 - 6 | 441 | 72 | ||
FQ232695.1 | - | 5 - 6 | 433 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |