Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077201.1 | NP_001070669.1 | 4 - 5 | 502 | 95 | |
XM_006249243.2 | XP_006249305.1 | 4 - 5 | 469 | 95 | ||
GenBank mRNA | DQ914829.1 | - | 3 - 4 | 376 | 95 | |
DQ914830.1 | - | 4 - 5 | 502 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |