Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009964.1 | NP_001009964.1 | 4 - 5 | 526 | 104 | |
XM_006249387.2 | XP_006249449.1 | 5 - 6 | 1021 | 104 | ||
XM_006249388.2 | XP_006249450.1 | 5 - 6 | 1150 | 104 | ||
XM_006249389.2 | XP_006249451.1 | 6 - 7 | 1325 | 104 | ||
XM_006249390.2 | XP_006249452.1 | 6 - 7 | 1196 | 104 | ||
XM_006249391.2 | XP_006249453.1 | 5 - 6 | 1170 | 104 | ||
XM_006249392.2 | XP_006249454.1 | 5 - 6 | 715 | 104 | ||
XM_008769230.1 | XP_008767452.1 | 6 - 7 | 1173 | 104 | ||
XM_008769231.1 | XP_008767453.1 | 5 - 6 | 826 | 104 | ||
GenBank mRNA | AY859491.1 | - | 4 - 5 | 526 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |