Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253357.2 | XP_006253419.1 | 3 - 4 | 2641 | 87 | |
GenBank mRNA | BC162035.1 | - | 2 - 3 | 1094 | 7097232 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |