Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057100.2 | NP_476441.2 | 5 - 6 | 594 | 91 | |
GenBank mRNA | BC070881.1 | - | 5 - 6 | 594 | 7096665 | 91 |
D42148.1 | - | 5 - 6 | 489 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |