Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106091.1 | NP_001099561.1 | 5 - 6 | 834 | 93 | |
XM_006253349.2 | XP_006253411.1 | 2055 | 93 | |||
GenBank mRNA | BC158691.1 | - | 5 - 6 | 820 | 9021908 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |