Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020302.1 | NP_064698.1 | 9 - 10 | 951 | 152 | |
GenBank mRNA | BC078839.1 | - | 9 - 10 | 857 | 7114315 | 152 |
Y07903.1 | - | 9 - 10 | 951 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |