Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adprhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013054.1 | NP_001013072.1 | 5 - 6 | 812 | 123 | |
XM_006253439.2 | XP_006253501.1 | 5 - 6 | 1286 | 123 | ||
GenBank mRNA | BC083773.1 | - | 5 - 6 | 812 | 7193406 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |