Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253451.2 | XP_006253513.1 | 2 - 3 | 324 | 107 | |
XM_008771397.1 | XP_008769619.1 | 3 - 4 | 327 | 107 | ||
XM_008771398.1 | XP_008769620.1 | 3 - 4 | 406 | 107 | ||
XM_008771399.1 | XP_008769621.1 | 3 - 4 | 234 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus Probe spans exons |