Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138532.1 | NP_612541.1 | 4 - 5 | 396 | 85 | |
XM_008769600.1 | XP_008767822.1 | 2 - 3 | 359 | 85 | ||
XM_008769601.1 | XP_008767823.1 | 2 - 3 | 361 | 85 | ||
GenBank mRNA | AF202049.1 | - | 4 - 5 | 396 | 85 | |
BC060314.1 | - | 4 - 5 | 417 | 6918381 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |