Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105952.1 | NP_001099422.1 | 2 - 3 | 537 | 138 | |
XM_006249762.2 | XP_006249824.1 | 1 - 2 | 198 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |