Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uchl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012149.1 | NP_001012149.1 | 4 - 5 | 460 | 125 | |
XM_006249960.2 | XP_006250022.1 | 4 - 5 | 495 | 125 | ||
XM_006249961.2 | XP_006250023.1 | 4 - 5 | 495 | 125 | ||
XM_006249962.2 | XP_006250024.1 | 4 - 5 | 496 | 125 | ||
XM_006249963.2 | XP_006250025.1 | 4 - 5 | 497 | 125 | ||
GenBank mRNA | BC088841.1 | - | 4 - 5 | 460 | 7125859 | 125 |
FQ220301.1 | - | 4 - 5 | 466 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |