Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sell | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019177.3 | NP_062050.3 | 3 - 4 | 583 | 63 | |
GenBank mRNA | D10831.1 | - | 3 - 4 | 575 | 63 | |
S79523.1 | - | 3 - 4 | 583 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |