Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250119.2 | XP_006250181.1 | 2 - 3 | 599 | 134 | |
XM_006250122.2 | XP_006250184.1 | 2 - 3 | 594 | 134 | ||
XM_006250123.2 | XP_006250185.1 | 2 - 3 | 590 | 134 | ||
XM_008769703.1 | XP_008767925.1 | 4 - 5 | 655 | 134 | ||
XM_008769704.1 | XP_008767926.1 | 2 - 3 | 591 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |