Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005871.1 | NP_001005871.1 | 11 - 12 | 2203 | 85 | |
XM_008769446.1 | XP_008767668.1 | 12 - 13 | 2586 | 85 | ||
XM_008769447.1 | XP_008767669.1 | 12 - 13 | 3223 | 85 | ||
XM_008769448.1 | XP_008767670.1 | 11 - 12 | 2550 | 85 | ||
XM_008769449.1 | XP_008767671.1 | 12 - 13 | 2587 | 85 | ||
XM_008769450.1 | XP_008767672.1 | 11 - 12 | 2551 | 85 | ||
GenBank mRNA | U15408.1 | - | 11 - 12 | 2203 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Rattus norvegicus Probe spans exons |