Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005871.1 | NP_001005871.1 | 6 - 7 | 1109 | 132 | |
XM_008769446.1 | XP_008767668.1 | 7 - 8 | 1492 | 132 | ||
XM_008769447.1 | XP_008767669.1 | 7 - 8 | 2129 | 132 | ||
XM_008769449.1 | XP_008767671.1 | 7 - 8 | 1493 | 132 | ||
GenBank mRNA | U15408.1 | - | 6 - 7 | 1109 | 132 | |
X76452.1 | - | 2 - 3 | 148 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Transporter Rattus norvegicus Probe spans exons |