Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769739.1 | XP_008767961.1 | 2 - 3 | 151 | 92 | |
GenBank mRNA | BC166877.1 | - | 1 - 2 | 93 | 6921627 | 92 |
EF158003.1 | - | 1 - 2 | 97 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |