Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025126.1 | NP_001020297.1 | 4 - 5 | 571 | 95 | |
XM_006250113.2 | XP_006250175.1 | 11 - 12 | 1770 | 95 | ||
XM_006250114.2 | XP_006250176.1 | 11 - 12 | 1758 | 95 | ||
XM_006250115.2 | XP_006250177.1 | 10 - 11 | 1710 | 95 | ||
XM_006250116.2 | XP_006250178.1 | 10 - 11 | 1698 | 95 | ||
GenBank mRNA | BC091284.1 | - | 4 - 5 | 571 | 7304516 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |