Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012025.1 | NP_001012025.1 | 4 - 5 | 620 | 115 | |
XM_006249699.2 | XP_006249761.1 | 4 - 5 | 663 | 115 | ||
GenBank mRNA | BC088845.1 | - | 4 - 5 | 620 | 7307995 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |