Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130409.2 | NP_569093.2 | 15 - 16 | 2453 | 99 | |
XM_008769531.1 | XP_008767753.1 | 14 - 15 | 2479 | 99 | ||
GenBank mRNA | AJ320522.1 | - | 2427 | 99 | ||
CK359694.1 | - | 2 - 3 | 334 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |