Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qsox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109898.1 | NP_001103368.1 | 2 - 3 | 406 | 86 | |
NM_053431.3 | NP_445883.1 | 2 - 3 | 406 | 86 | ||
GenBank mRNA | AB044285.1 | - | 2 - 3 | 406 | 86 | |
AF217799.1 | - | 2 - 3 | 410 | 86 | ||
AF285078.1 | - | 2 - 3 | 428 | 86 | ||
AY623665.1 | - | 2 - 3 | 408 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |