Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107173.2 | NP_001100643.1 | 4 - 5 | 1368 | 110 | |
XM_006249770.2 | XP_006249832.1 | 3 - 4 | 1460 | 110 | ||
XM_006249771.2 | XP_006249833.1 | 3 - 4 | 1156 | 110 | ||
XM_006249772.2 | XP_006249834.1 | 3 - 4 | 1289 | 110 | ||
XM_006249773.2 | XP_006249835.1 | 3 - 4 | 1480 | 110 | ||
XM_008769467.1 | XP_008767689.1 | 4 - 5 | 1733 | 110 | ||
GenBank mRNA | BC168230.1 | - | 3 - 4 | 1384 | 7382659 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |