Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144561.2 | NP_653147.1 | 6 - 7 | 946 | 118 | |
NM_144562.1 | NP_653148.1 | 4 - 5 | 551 | 118 | ||
XM_006250144.2 | XP_006250206.1 | 6 - 7 | 881 | 118 | ||
GenBank mRNA | AF458416.1 | - | 6 - 7 | 947 | 118 | |
AF458417.1 | - | 4 - 5 | 551 | 118 | ||
BC081721.1 | - | 6 - 7 | 936 | 7107835 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |